[EN]20 COMPONENTS MIX 1 * 30 mL
Catalog # CMPLRD101282L03.6
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
[EN]20 COMPONENTS MIX 1 * 30 mL
Catalog # CMPLRD101282L03.6
Supplier: CUSTOM MADE CHEMICALS LAB
New Product
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
pH buffer solutions
Supplier: Mettler - Toledo
VWR® AVS TITRINORM, pufrovací roztoky, 20 °C
Supplier: VWR Chemicals
Roztoky standardní pro měření vodivosti
Supplier: VWR Chemicals
Roztoky pufrů v sáčcích, Certipur®, Supelco®
Supplier: Merck
Chlorid draselný 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
VWR®, Conductivity standards at 25 °C, potassium chloride solutions
Supplier: VWR Chemicals
pH Standard Solutions, IUPAC, Certified
Supplier: Hach