Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
PI Brite™

PI Brite™

2070, 1 mg
Katalogové číslo AATB17583
Dodavatel:   AAT Bioquest
PI Brite™
Katalogové číslo AATB17583
Dodavatel:  AAT Bioquest
Číslo CAS:  
PI Brite™

2070, 1 mg

Přejít na:
Číslo CAS:  
Price
3 490,00 Kč
Přihlásit se pro dostupnost
1 * 1 mg
Product Details & Documents

PI Brite™ is a propidium iodide (PI) analog developed by AAT Bioquest to improve the brightness and fixability of propidium iodide in cells.

  • Stable for 24 months

Quality tested in-house

Caution: For Research Use Only

  • Molecular Weight:1178.51 g/mol
Specifikace

Specifikace

  • Bal.:
    1 mg
  • Sekvence:
    2070

Specifikace

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.