Zadání objednávky
Czech Republic
Orders LinkContactUsLinkComponent
 

PI Brite™

2210, 5 mg
  AATB17584
PI Brite™
  AATB17584
 :  
PI Brite™

2210, 5 mg

 :  
 
7 120,00 Kč
   
1 * 5 mg
 

PI Brite™ is a propidium iodide (PI) analog developed by AAT Bioquest to improve the brightness and fixability of propidium iodide in cells.

  • Stable for 24 months

Quality tested in-house

 : For Research Use Only

  •  :1178.51 g/mol
 

 

  • Bal.:
    5 mg
  • Sekvence:
    2210