Urine Cups with Integrated Transfer Device
The Urine Cup with integrated transfer device allows you to collect and transport urine samples safely to the necessary destination. Thanks to the integrated transfer device, sample transfer from the cup to an evacuated tube is simple, fast and hygienic.
- Recessed transfer device to reduce risk of needle stick injuries
- Available bulk packed or in cases of small packs to suit different distribution needs within healthcare facilities
The optimised threading of the urine cup makes it easy to open and close. In addition, a stabilising flange makes the cup closure more secure and prevents over-threading.
The perforated label acts as a sterility integrity seal visually indicating if the product has been opened prior to use.
- Typ uzávěru:Šroubovací víčko
- Sterilita:Sterile
- Objem:100 ml
- Popis:Urine cup with integrated transfer device
- Sekvence:01
- Barva víčka:Žlutá