Orders LinkContactUsLinkComponent
Racks for test tubes
Racks for test tubes
03, 24 mm, —, PE
Catalog # HACH1488600
Supplier:   Hach
Racks for test tubes
Catalog # HACH1488600
Supplier:  Hach
Racks for test tubes
Racks for test tubes
03, 24 mm, —, PE
Price
779,00 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
These test tube racks features a special blend of polyethylene which sinks in water, assuring non-floating racks when immersed.
  • Stackable
  • Good chemical resistance
  • Good temperature resistance
Specifications
  • Ø otvoru:
    24 mm
  • Poč. otvorů:
  • Materiál:
    PE
  • Sekvence:
    03