MUC1 REC MAB SM3 RABBIT IGGΛ 1 * 200 µG
Catalog # ENZOENZABS6460200
Supplier: Enzo Life Sciences
Frequently Bought Together
VWR®, Square Bottles, PET
Supplier: LAB3 OWN PRODUCTION
Cell culture plates, 384-well, CELLSTAR®
Supplier: Greiner Bio-One
Racks and lids for storage 2D barcoded tubes, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
Spinner flasks, disposable, Corning®
Supplier: Corning
Tissue vial box, blue, WHEATON® CryoFILE
Supplier: DWK Life Sciences
Cell culture vessels, Corning® HYPERflask™ M, CellBIND® treated
Supplier: Corning
Storage tubes, 2D barcoded, Matrix™
Supplier: THERMO MATRIX TECHNOLOGIES
Micro packaging vials with thread, Nalgene®
Supplier: Thermo Fisher Scientific
Racks for culture tubes, stainless steel
Supplier: BELLCO GLASS
Test tubes with screw caps
Supplier: BOETTGER PAUL
[EN]LID FOR ICE PAN 9 L-GRN 1 * 1 KS
Supplier: VWR Collection
Storage Boxes, PP, with Lid
Supplier: KOEHLER TECHNISCHE PRODUKTEN
Centrifuge tubes, PP, with screw cap
Supplier: KARTELL
Corning® Gosselin™ Petri Dish, Aseptic
Supplier: Corning