MUC1 REC MAB HMFG2 MS IGG1Λ 1 * 200 µG
ENZOENZABS6430200
Frequently Bought Together
Sealing film, Axygen® AxySeal
: Corning
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
: VWR Collection
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
: VWR Collection
VWR®, Pipety Pasteurovy, Standard Line
: VWR Collection
VWR®, Nádoby na vzorky se šroubovacím uzávěrem
: VWR Collection
VWR®, Pipety Pasteurovy, High performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Kryoboxy a pořadače
: VWR Collection