P-GLYCOPROTEIN REC MAB MRK16 MS IGG1Κ 1 * 100 µG
ENZOENZABS5580100
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
: EPPENDORF
VWR® Premium Line, Pipety sérologické
: VWR Collection
VWR®, Pipety Pasteurovy, Standard Line
: VWR Collection
Transfer pipettes
: RATIOLAB
VWR®, Cell Culture Flasks, Non Treated or Treated
: VWR Collection
VWR®, Nádoby na vzorky se šroubovacím uzávěrem
: VWR Collection
VWR®, Zkumavky pro PCR, 0,2 ml
: VWR Collection
VWR®, Sérologická pipeta, standardní
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
: VWR Collection
VWR®, Roztěrky, tvar L, jednorázové
: VWR Collection
VWR®, Pipety Pasteurovy, High performance
: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF