FLUORESCEIN RAB (4-4-20) (MS IGG2C) 1 * 200 µG
Katalogové číslo ENZOENZABS5050200
Dodavatel: ENZO LIFE SCIENCES
FLUORESCEIN RAB (4-4-20) (MS IGG2C) 1 * 200 µG
Katalogové číslo ENZOENZABS5050200
Dodavatel: ENZO LIFE SCIENCES
Frequently Bought Together
VWR®, Misky kultivační, vícejamkové
Dodavatel: VWR Collection
Petri dishes
Dodavatel: GAMA GROUP
VWR®, Centrifuge Tubes, Conical Bottom
Dodavatel: VWR Collection
VWR®, Sérologická pipeta, standardní
Dodavatel: VWR Collection
Centrifuge tubes, 15 ml, Corning®
Dodavatel: Corning
VWR®, Misky Petriho, PS
Dodavatel: VWR Collection
VWR®, Cell Culture Flasks, Non Treated or Treated
Dodavatel: VWR Collection
VWR®, Centrifuge Tubes, High Performance, Flat or Plug Cap, PP
Dodavatel: VWR Collection
Multiwell cell culture plates, Falcon®
Dodavatel: Corning
VWR®, Pipety Pasteurovy, Standard Line
Dodavatel: VWR Collection
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Dodavatel: EPPENDORF
VWR® Ultra-High Performance Centrifuge Tubes
Dodavatel: VWR Collection
Cryoboxes, Nalgene®
Dodavatel: Thermo Fisher Scientific
VWR®, Proužky PCR zkumavek a uzávěrů
Dodavatel: VWR Collection
VWR®, Pipety Pasteurovy, High performance
Dodavatel: VWR Collection
VWR®, Kryoboxy, lepenka, 133×133 mm
Dodavatel: VWR Collection
Kryoboxy, 133×133 mm
Dodavatel: TENAK
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Dodavatel: VWR Collection