IL-22 HU REC 1 * 100 µG
Catalog # ENZOENZPRT2500100
Supplier: Enzo Life Sciences
Frequently Bought Together
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Supplier: BIOCISION
Cryo vial closure colour coders, Nalgene®
Supplier: Thermo Fisher Scientific
Kryoboxy, 133×133 mm
Supplier: TENAK
VWR®, Cryo coder cap inserts
Supplier: VWR Collection
VWR®, Kryoboxy, lepenka, 133×133 mm
Supplier: VWR Collection
Immunoassay microplates
Supplier: Thermo Fisher Scientific
Petri dishes, contact plates
Supplier: Anicrin
Screw caps for T501 sample tubes
Supplier: Simport Scientific
Eppendorf® Microplates
Supplier: EPPENDORF
Volumetric measuring scoops, SteriWare®
Supplier: Sampling Systems
Poly-D-Lysine flasks, Corning® BioCoat®
Supplier: Corning
Microplates, 96-well
Supplier: RATIOLAB