FGF-9 MS REC 1 * 100 µG
Katalogové číslo ENZOENZPRT2330100
Dodavatel: Enzo Life Sciences
Frequently Bought Together
Cryoboxes, B69/B50/B80/B99-Series
Dodavatel: GLW
Colour coders for cryovials, CryoColorCode, Nunc™
Dodavatel: Thermo Fisher Scientific
[EN]MICRORACK TUBES - 1.2ML -PP- 1 * 1.000 KS
Dodavatel: Greiner Bio-One
Mikrocentrifugační zkumavky, 0,5/1,5/2,0 ml
Dodavatel: Brand
Ice pans, Bel-Art Magic Touch 2™
Dodavatel: Bel-Art Products, a Part of SP
Caps for microtubes
Dodavatel: RATIOLAB
Microtube racks
Dodavatel: RATIOLAB
Microcentrifuge tubes, 0,5/0,7/1,5/2,0/2,2 ml
Dodavatel: Greiner Bio-One
Deepwell Plates 96-well, 1000 µl
Dodavatel: EPPENDORF
Sterilin™ Samples Containers with Screw Caps, PS, 60 to 250 ml
Dodavatel: Sterilin
Storage rack for bags, BagRack® <i>Slide</i>
Dodavatel: INTERSCIENCE
Systém AriaMx Real-Time PCR
Dodavatel: AGILENT