FGF7 HU REC 1 * 10 µG
Katalogové číslo ENZOENZPRT2020010
Dodavatel: ENZO LIFE SCIENCES
Frequently Bought Together
VWR® Standard Line, Láhve úzkohrdlé, se šroubovacím uzávěrem
Dodavatel: VWR Collection
VWR®, Proužky PCR zkumavek a uzávěrů
Dodavatel: VWR Collection
Colour coders for cryovials, CryoColorCode, Nunc™
Dodavatel: Thermo Fisher Scientific
VWR® Cryoboxes, Cardboard, 136×136 mm, with Full Labelling
Dodavatel: VWR Collection
Cell culture plates and Multidishes, Nunclon™ Sphera™
Dodavatel: Thermo Fisher Scientific
MicroWell™ Plates, 96 well, Nunc-Immuno™
Dodavatel: Thermo Fisher Scientific
Microcentrifuge tubes, ClikLok™ and MacrewTube™
Dodavatel: Simport Scientific
Sample containers, with screw cap, STERILIN®
Dodavatel: Sterilin
Screw cap microcentrifuge tubes, SuperClear®
Dodavatel: LABCON
Storage tubes, 2D barcoded, Matrix™
Dodavatel: THERMO MATRIX TECHNOLOGIES
Sample container, with screw cap
Dodavatel: DELTALAB
Multiple well cell culture plates, Costar®
Dodavatel: Corning
Containers with snap caps, Corning® Gosselin™
Dodavatel: Corning
384-well clear polystyrene microplates, Corning®
Dodavatel: Corning