SONIDEGIB FREE BASE 1 * 25 mg
Catalog # ENZOENZCHM2380025
Supplier: ENZO LIFE SCIENCES
Frequently Bought Together
Hydroxid sodný, pellets, EMSURE® p.a., Supelco®
Supplier: Merck
Kyselina trifluoroctová ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Giemsa's stain solution for microscopy, Sigma-Aldrich®
Supplier: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Supplier: Merck
Formaldehyde 4% stabilised, buffered (pH 7,0 ±0,1)
Supplier: VWR Chemicals
Chlornan sodný (3,5% Cl₂) in aqueous solution, GPR RECTAPUR®
Supplier: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Supplier: Merck
Hydroxid sodný, pellets, pure, Supelco®
Supplier: Merck
Kyselina mravenčí ≥99%, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
VWR®, Práškové balíčky pro stanovení chlóru, činidla pro fotometry
Supplier: VWR Chemicals
Test strips, peroxide, Dosatest
Supplier: VWR Chemicals
VWR® Dosatest, pH indicator test strips, non bleeding
Supplier: VWR Chemicals
Accessories
Supplier: Hach
Test kits, photometric, COD, LCK cuvette tests
Supplier: Hach
[EN]BIOTECTOR 1.8N SULFURIC ACID 80MG/L 1 * 20 L
Supplier: Hach