ALECTINIB HYDROCHLORIDE 1 * 200 mg
Katalogové číslo ENZOENZCHM2130200
Dodavatel: ENZO LIFE SCIENCES
Frequently Bought Together
Kyselina trifluoroctová ≥99.9% for peptide synthesis
Dodavatel: VWR Chemicals
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
n-Pentan, EMSURE® p.a., Supelco®
Dodavatel: Merck
Aceton
Dodavatel: Lach-ner S.R.O.
Dichlormethan ≥99.9% (by GC) stabilised, Puriss. p.a. ACS, ISO analytical reagent, Riedel-de Haën™
Dodavatel: Honeywell Chemicals
Ethylacetát ≥99.5% (by GC), Puriss. p.a. ACS, ISO, Reag. Ph. Eur., Riedel-de Haën™
Dodavatel: Honeywell Chemicals
PETROLEUM ETHER 40-60C FOR PESTICIDE AN 1 * 2,5 L
Dodavatel: PanReac AppliChem
Petrolether, 30…40 °C, AnalaR NORMAPUR® analytical reagent
Dodavatel: VWR Chemicals
Tetrahydrofuran ≥99.9% stabilizovaný, Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Dodavatel: Honeywell Chemicals
Ethylacetát, extra dry over Molecular Sieve 99.9%, AcroSeal®
Dodavatel: Thermo Fisher Scientific
Bromoform 99+% stabilizovaný
Dodavatel: Thermo Fisher Scientific
L-LYSINE MONOHYDROCHLORIDE (PH. EUR US 1 * 25 kg
Dodavatel: PanReac AppliChem
Voda, CHROMASOLV™ for HPLC, Riedel-de Haën™
Dodavatel: Honeywell Chemicals
ISOAMYL ALCOHOL ACCORDING TO GERBER FOR 1 * 1.000 mL
Dodavatel: PanReac AppliChem