ALECTINIB HYDROCHLORIDE 1 * 25 mg
Katalogové číslo ENZOENZCHM2130025
Dodavatel: ENZO LIFE SCIENCES
Frequently Bought Together
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Aceton ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
n-Hexan ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Methanol, anhydrous (max. 0.003% H₂O) analytical reagent
Dodavatel: Merck
Ethanol 96% vol Ph. Eur.
Dodavatel: VWR Chemicals
Voda, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Dodavatel: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Dodavatel: VWR Chemicals
n-Heptan, EMPLURA®, Supelco®
Dodavatel: Merck
Acetonitril, LiChrosolv®, hypergrade for LC-MS, Supelco®
Dodavatel: Merck
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Chloroform, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
Dichlormethan stabilised, LiChrosolv® for HPLC, Supelco®
Dodavatel: Merck
Acetonitrile : water : trifluoroacetic acid (50 : 47,5 : 2,5), HiPerSolv CHROMANORM®
Dodavatel: VWR Chemicals