COLLAGEN TYPE V MAB 1E2-E4/COL5 1 * 400 µG
Katalogové číslo ENZOBPDCSI0060104
Dodavatel: ENZO LIFE SCIENCES
COLLAGEN TYPE V MAB 1E2-E4/COL5 1 * 400 µG
Katalogové číslo ENZOBPDCSI0060104
Dodavatel: ENZO LIFE SCIENCES
Frequently Bought Together
Eppendorf Tubes® 3810X Microtubes 1,5 ml
Dodavatel: EPPENDORF
VWR® Premium Line, Pipety sérologické
Dodavatel: VWR Collection
VWR® Inoculating Loops and Needles, Disposable
Dodavatel: VWR Collection
VWR®, Centrifuge Tubes, Freestanding, with Flat Screw Caps
Dodavatel: VWR Collection
VWR®, Sérologická pipeta, standardní
Dodavatel: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
Dodavatel: VWR Collection
Centrifuge tubes, conical bottom, Falcon®
Dodavatel: Corning
VWR® Water Sampling Bottles, with Tamper Evident Cap
Dodavatel: VWR Collection
Cell culture dishes, Falcon®
Dodavatel: Corning
VWR®, Cell Culture Flasks, Non Treated or Treated
Dodavatel: VWR Collection
VWR®, Nádoby na vzorky se šroubovacím uzávěrem
Dodavatel: VWR Collection
Transfer pipettes
Dodavatel: RATIOLAB
VWR®, Pipety Pasteurovy, Standard Line
Dodavatel: VWR Collection
VWR®, Pipety Pasteurovy, High performance
Dodavatel: VWR Collection
Safe-Lock Tubes, 1,5 ml - Microtubes
Dodavatel: EPPENDORF
VWR®, Kryoboxy, lepenka, 133×133 mm
Dodavatel: VWR Collection