Orders LinkContactUsLinkComponent
Cell counting kit-8
Cell counting kit-8
04, Cell counting kit-8, 500 Tests
Katalogové číslo ENZOALX850039KI02
Dodavatel:   Enzo Life Sciences
Cell counting kit-8
Katalogové číslo ENZOALX850039KI02
Dodavatel:  Enzo Life Sciences
Číslo CAS:  
Cell counting kit-8
Cell counting kit-8
04, Cell counting kit-8, 500 Tests
Přejít na:
Číslo CAS:  
Price
16 530,00 Kč
Přihlásit se pro dostupnost
1 * 500 Tests
Product Details & Documents
Colorimetric kit for simple and accurate cell proliferation and cytotoxicity assays. Advantages: One-step, ready-to-use solution, no radioisotopes, no solubilization step, high sensitivity, correlates with the [3H]-thymidine incorporation assay. Can be added directly to the cell media for fast, high-throughput screening without a solubilization process obtaining highly reproducible and accurate results.
Informace pro objednávání: Quantity: 500 tests/5ml; 2500 tests/5 x 5ml. Contents: 5mM WST-8 solution, 0.2mM 1-methoxy-PMS, 150mM sodium chloride.
Specifikace
Specifikace
  • Bal.:
    500 Tests
  • Popis:
    Cell counting kit-8
  • Sekvence:
    04
  • Cat. No.:
    ENZOALX850039KI02
  • Rozměry:
    5×500 tests