X-Gal
3173, Glass bottle for solids, 5 g
Katalogové číslo ENZOALX582002G005
Dodavatel: Enzo Life Sciences
Přejít na:
Product Details & Documents
Synonyma: BCIG, X-GAL, (2S,3R,4S,5R,6R)-2-((5-Bromo-4-chloro-1H-indol-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol
- Číslo CAS:7240-90-6
- Vzorec:C₁₄H₁₅BrClNO₆
- MDL číslo:MFCD00005666
- Molecular Weight:408.63
- Bod tání:237…239 °C
- Teplota pro skladování:Freezer
- EINECS:230-640-8
Specifikace
Specifikace
- Typ balení:Glass bottle for solids
- Bal.:5 g
- Sekvence:3173
Specifikace
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



