IL-6 MS REC 1 * 100 µG
Katalogové číslo ENZOALX2018190100
Dodavatel: ENZO LIFE SCIENCES
Frequently Bought Together
Safe-Lock Tubes, 2,0 ml - Microtubes
Dodavatel: EPPENDORF
VWR®, Centrifuge Tubes
Dodavatel: VWR Collection
VWR® Media Bottles, Square, PETG
Dodavatel: VWR Collection
CryoBoxes with hinged lid, for cryovials, 1 to 5 ml, TruCool®
Dodavatel: BIOCISION
VWR® Ultra-High Performance Centrifuge Tubes
Dodavatel: VWR Collection
96-Well cell culture plates, Falcon®
Dodavatel: Corning
VWR®, Pipety Pasteurovy, High performance
Dodavatel: VWR Collection
Petri dishes
Dodavatel: GAMA GROUP
VWR®, Sérologická pipeta, standardní
Dodavatel: VWR Collection
Round-bottom polystyrene tubes, disposable, Falcon®
Dodavatel: Corning
VWR®, Centrifuge Tubes, Metal-Free, with Screw Cap
Dodavatel: VWR Collection
Stoppers, press down, VacuCap®
Dodavatel: Simport Scientific
VWR®, Pipety Pasteurovy, Standard Line
Dodavatel: VWR Collection
Seals for ALPS™ 50 V heat sealer
Dodavatel: THERMO MATRIX TECHNOLOGIES
VWR®, Vzorkovací lžíce
Dodavatel: VWR Collection
Centrifuge tubes with flat caps, MetalFree®
Dodavatel: LABCON
96-Well deep well blocks
Dodavatel: HAMILTON ROBOTICS
Serological pipettes
Dodavatel: Greiner Bio-One