Micro forceps
06, No. 1, straight, anti-magnetic, very fine tips, 120 mm
Katalogové číslo HAMMHSC607-12
Dodavatel: HAMMACHER KARL
Přejít na:
Product Details & Documents
Stainless steel.
Specifikace
- Type:No. 1, straight, anti-magnetic, very fine tips
- Délka:120 mm
- Sekvence:06