[EN]8 COMPOSANTS DANS L'EAU 1 * 100 mL
Katalogové číslo CMPLRD094793L1
Dodavatel: CUSTOM MADE CHEMICALS LAB
[EN]8 COMPOSANTS DANS L'EAU 1 * 100 mL
Katalogové číslo CMPLRD094793L1
Dodavatel: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Kyselina octová 0.1 mol/l (0.1 N), AVS TITRINORM volumetric solution
Dodavatel: VWR Chemicals
EOSIN RAL 555 1 * 1 L
Dodavatel: RAL
Test strips for copper, MQuant®, Supelco®
Dodavatel: Merck
Hydroxid sodný 35% in aqueous solution, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Hydroxid sodný 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Dodavatel: VWR Chemicals
Hydroxid vápenatý, EMSURE® ACS, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
Test strips, peracetic acid, QUANTOFIX®
Dodavatel: MACHEREY-NAGEL
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Dodavatel: VWR Chemicals
Test pro stanovení peroxidu, MQuant™, Supelco®
Dodavatel: Merck
Kyselina 1-oktansulfonová sodná sůl ≥98%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Ethanol 70% ≥70% (v/v)
Dodavatel: VWR Chemicals
Test kits, ammonium, Spectroquant®, Supelco®
Dodavatel: Merck
BOND EPITOPE RETRIEVAL SOLUTION 2 1 * 1 L
Dodavatel: LEICA MICROSYSTEMS
Test kits, photometric, iron, LCK cuvette tests
Dodavatel: Hach
Test kits, photometric, COD, LCK cuvette tests
Dodavatel: Hach