Orders LinkContactUsLinkComponent
Viscometers, ROTAVISC SBS
Viscometers, ROTAVISC SBS
05, Rotational viscometer package, EU plug, ROTAVISC SBS me-vi HELI Complete, 100 - 40000000 mPas
Katalogové číslo 531-1478
Dodavatel:   IKA
Viscometers, ROTAVISC SBS
Katalogové číslo 531-1478
Dodavatel:  IKA
Viscometers, ROTAVISC SBS
Viscometers, ROTAVISC SBS
05, Rotational viscometer package, EU plug, ROTAVISC SBS me-vi HELI Complete, 100 - 40000000 mPas
Price
124 120,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
SBS (Steel Ball Bearing, Suspended) rotational viscometers extend a large part of the application spectrum of the IKA ROTAVISC device family.
  • Easy to use
  • Highest measurement accuracy
  • Infinitely variable speed
Instead of jewel bearing, these devices are equipped with a special ball bearing technology.
They are characterised by their high robustness. Predestined for applications with very high sample throughput, frequent spindle changes, operation in harsher environments, or even frequently changing device locations.
Whether simple or demanding viscosity measurements - with ROTAVISC you get fast and accurate results. Intuitive menu navigation awaits you via the generous 4,3" TFT display. Digital levelling also supports the correct setting of the starting condition. Recurring tasks are facilitated by the simple ramp function.
The accuracy of the ROTAVISC rotary viscometers for Newtonian and non Newtonian liquids is ±1 % of the measuring range. Reproducibility is ±0,2%. With ROTAVISC you have a viscometer in which you can adjust the speed steplessly, but also specify programs or individual speed levels.
Informace o dodávce: Complete packages include ROTAVISC SBS (hi-vi I, hi-vi II, or me-vi control), ROTASTAND, SP set-2, PT 100.8, and Guard rail.
HELI complete packages include ROTAVISC SBS (hi-vi I, hi-vi II, or me-vi control), HELISTAND, T-SP-SET, SP set-2, PT 100.8, and Guard rail.
Specifikace
  • Model:
    ROTAVISC SBS me-vi HELI Complete
  • Rozsah:
    100 - 40000000 mPas
  • Popis:
    Rotational viscometer package, EU plug
  • Sekvence:
    05
  • Hmotnost:
    7,1 kg
  • Cat. No.:
    531-1478
  • Displej:
    TFT
  • Š×H×V:
    351×372×629 mm
Accessories
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Stand for ROTAVISC
    Stand for ROTAVISC
    IKAA20116428
    14
    Přihlásit se pro dostupnost
    1 * 1 KS - 26 440,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Standardní silikonový olej, CAL-O-100
    Standardní silikonový olej, CAL-O-100
    IKAA25000434
    04
    Přihlásit se pro dostupnost
    1 * 500 mL - 4 290,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Standard silicone oil, CAL-O-30000
    Standard silicone oil, CAL-O-30000
    IKAA25000467
    09
    Přihlásit se pro dostupnost
    1 * 500 mL - 4 290,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Standardní silikonový olej, CAL-O-1000
    Standardní silikonový olej, CAL-O-1000
    IKAA25000436
    06
    Přihlásit se pro dostupnost
    1 * 500 mL - 4 290,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Standard silicone oil, CAL-O-60000
    Standard silicone oil, CAL-O-60000
    IKAA25000468
    10
    Přihlásit se pro dostupnost
    1 * 500 mL - 4 290,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • HELISTAND set includes T-spindles for non-flowing samples, EU plug
    HELISTAND set includes T-spindles for non-flowing samples, EU plug
    IKAA20128647
    04
    Přihlásit se pro dostupnost
    1 * 1 KS - 46 910,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Teplotní rozsah
    Dostupnost
    Price
  • CBC VISC lite temperature control, EU plug
    CBC VISC lite temperature control, EU plug
    IKAA20102007
    01
    −25...+125 °C
    Přihlásit se pro dostupnost
    1 * 1 KS - 104 780,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Adaptér VOLS-1 pro měření viskozity zvláště cenných a drahých vzorků
    Adaptér VOLS-1 pro měření viskozity zvláště cenných a drahých vzorků
    IKAA25000352
    03
    Přihlásit se pro dostupnost
    1 * 1 SET - 37 150,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • VOL-SP-7.1, spindle for extended viscosity range, suitable for sample volumes of 7,1 ml
    VOL-SP-7.1, spindle for extended viscosity range, suitable for sample volumes of 7,1 ml
    IKAA25006849
    13
    Přihlásit se pro dostupnost
    1 * 1 KS - 10 280,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Standard silicone oil, CAL-O-5000
    Standard silicone oil, CAL-O-5000
    IKAA25000437
    07
    Přihlásit se pro dostupnost
    1 * 500 mL - 4 290,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Standardní silikonový olej, CAL-O-5
    Standardní silikonový olej, CAL-O-5
    IKAA25000397
    01
    Přihlásit se pro dostupnost
    1 * 500 mL - 4 290,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • VOL-SP-11, spindle for extended viscosity range, suitable for sample volumes of 11 ml
    VOL-SP-11, spindle for extended viscosity range, suitable for sample volumes of 11 ml
    IKAA25006851
    11
    Přihlásit se pro dostupnost
    1 * 1 KS - 10 580,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Standard silicone oil, CAL-O-12500
    Standard silicone oil, CAL-O-12500
    IKAA25000466
    08
    Přihlásit se pro dostupnost
    1 * 500 mL - 4 290,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Standardní silikonový olej, CAL-O-500
    Standardní silikonový olej, CAL-O-500
    IKAA25000435
    05
    Přihlásit se pro dostupnost
    1 * 500 mL - 4 290,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Labworldsoft® 6 Pro, with access to the entire device library
    Labworldsoft® 6 Pro, with access to the entire device library
    IKAA20017366
    02
    All configurations with any number of devices can be controlled at the same time
    Přihlásit se pro dostupnost
    1 * 1 KS - 336 600,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Adaptér ELVAS-1 pro měření vzorků s velmi nízkou viskozitou
    Adaptér ELVAS-1 pro měření vzorků s velmi nízkou viskozitou
    IKAA25000390
    02
    Přihlásit se pro dostupnost
    1 * 1 SET - 43 270,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • VOL-SP-13.5, spindle for extended viscosity range, suitable for sample volumes of 13,5 ml
    VOL-SP-13.5, spindle for extended viscosity range, suitable for sample volumes of 13,5 ml
    IKAA25006852
    10
    Přihlásit se pro dostupnost
    1 * 1 KS - 10 580,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Hose, silicone, 2×1,5 m, Ø 8 mm
    Hose, silicone, 2×1,5 m, Ø 8 mm
    IKAA4569000
    07
    Přihlásit se pro dostupnost
    1 * 1 KS - 1 820,00 Kč
Informace o produktu
  • Katalogové číslo
    Dostupnost
    Price
  • VOLS-1 flex adapter set for low volumes
    VOLS-1 flex adapter set for low volumes
    IKAA20114188
    Přihlásit se pro dostupnost
    1 * 1 KS - 18 910,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Standardní silikonový olej, CAL-O-10
    Standardní silikonový olej, CAL-O-10
    IKAA25000398
    02
    Přihlásit se pro dostupnost
    1 * 500 mL - 4 290,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Labworldsoft® 6 Starter
    Labworldsoft® 6 Starter
    IKAA20019397
    01
    Small configurations with up to 3 devices can be controlled at the same time
    Přihlásit se pro dostupnost
    1 * 1 KS - 147 530,00 Kč
Informace o produktu
  • Katalogové číslo
    Typ
    Tepl. stabilita
    Dostupnost
    Price
  • HRC 2 temperature control
    HRC 2 temperature control
    IKAA25004524
    HRC 2 control
    0,05±K
    Přihlásit se pro dostupnost
    1 * 1 KS - 171 840,00 Kč
Informace o produktu
  • Katalogové číslo
    Dostupnost
    Price
  • High-temperate system ROTATEMP, for ROTAVISC up to 300 °C, EU plug
    High-temperate system ROTATEMP, for ROTAVISC up to 300 °C, EU plug
    IKAA20113825
    Přihlásit se pro dostupnost
    1 * 1 KS - 104 560,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Adaptér, DINS-1, podle normy DIN 53019
    Adaptér, DINS-1, podle normy DIN 53019
    IKAA25000387
    01
    Přihlásit se pro dostupnost
    1 * 1 SET - 65 370,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Standard silicone oil, CAL-O-100000
    Standard silicone oil, CAL-O-100000
    IKAA25000469
    11
    Přihlásit se pro dostupnost
    1 * 500 mL - 4 290,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • VOL-SP-10.4, spindle for extended viscosity range, suitable for sample volumes of 10,4 ml
    VOL-SP-10.4, spindle for extended viscosity range, suitable for sample volumes of 10,4 ml
    IKAA25006850
    12
    Přihlásit se pro dostupnost
    1 * 1 KS - 10 580,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Určeno pro
    Dostupnost
    Price
  • Labworldsoft® 6 Visc
    Labworldsoft® 6 Visc
    IKAA20101872
    03
    Viscosity measurements for thermostats and pH meters of all manufacturers
    Přihlásit se pro dostupnost
    1 * 1 KS - 32 810,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Spirálový adaptér SAS-1, pro měření viskozity velmi lepivých a jiných viskózních vzorků
    Spirálový adaptér SAS-1, pro měření viskozity velmi lepivých a jiných viskózních vzorků
    IKAA25000377
    08
    Přihlásit se pro dostupnost
    1 * 1 SET - 34 510,00 Kč
Informace o produktu
  • Katalogové číslo
    Dostupnost
    Price
  • VOL-SP-2.1 Cylinder spindle with hook suspension, 95,8 mm
    VOL-SP-2.1 Cylinder spindle with hook suspension, 95,8 mm
    IKAA20118239
    Přihlásit se pro dostupnost
    1 * 1 KS - 10 580,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Sada křídlového vřetena (sada obsahuje 4 ks) pro měření viskozity suspenzí
    Sada křídlového vřetena (sada obsahuje 4 ks) pro měření viskozity suspenzí
    IKAA25000407
    09
    Přihlásit se pro dostupnost
    1 * 1 SET - 32 810,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Standardní silikonový olej, CAL-O-50
    Standardní silikonový olej, CAL-O-50
    IKAA25000399
    03
    Přihlásit se pro dostupnost
    1 * 500 mL - 4 290,00 Kč