Rhod-4™ AM
1442, Plastic bottle, 1.000 µG
Katalogové číslo AATB21123
Dodavatel: AAT Bioquest
Přejít na:
Product Details & Documents
Calcium measurement is critical for numerous biological investigations.
- Well-cited
- Long-wavelength emission
- Cell permeant
Caution: For research use only.
- Molecular Weight:1015.96 g/mol
- ADR:5,II
Specifikace
Specifikace
- Typ balení:Plastic bottle
- Bal.:1.000 µG
- Sekvence:1442



