Rhod-4™ AM
2070, Plastic bottle, 1 mg
Katalogové číslo AATB21120
Dodavatel: AAT Bioquest
Přejít na:
Product Details & Documents
Calcium measurement is critical for numerous biological investigations.
- Well-cited
- Long-wavelength emission
- Cell permeant
: For research use only.
- Molecular Weight:1015.96 g/mol
- ADR:5,II
Specifikace
Specifikace
- Typ balení:Plastic bottle
- Bal.:1 mg
- Sekvence:2070



