SPREJ KLERCIDE 70/30 IPA FILTROVANÝ 1L 1 * 6 KS
Katalogové číslo SHIE6600947
Dodavatel: ECOLAB CONTAMINATION CONTROL
SPREJ KLERCIDE 70/30 IPA FILTROVANÝ 1L 1 * 6 KS
Katalogové číslo SHIE6600947
Dodavatel: ECOLAB CONTAMINATION CONTROL
Frequently Bought Together
Diethylkarbonát for synthesis, Sigma-Aldrich®
Dodavatel: Merck
1-Acetyl-2-phenylhydrazine for synthesis, Sigma-Aldrich®
Dodavatel: Merck
2-Butoxyethanol 99%, extra pure
Dodavatel: Thermo Fisher Scientific
Chloroform ≥99.8% stabilizovaný, CHROMASOLV™ for HPLC, Riedel-de Haën™
Dodavatel: Honeywell Chemicals
1-Propanol ≥99.5% ACS
Dodavatel: Thermo Fisher Scientific
terc-Butylmethylether p.a.
Dodavatel: Thermo Fisher Scientific
2,4,6-Tris(2-pyridyl)-1,3,5-triazine, Supelco®
Dodavatel: Merck
Benzylalkohol 99-100.5% (by GC), Puriss., meets analytical specification of BP, Ph. Eur., NF, Riedel-de Haën™
Dodavatel: Honeywell Chemicals
Ethanol absolute (taxed) ≥99.8% (by GC)
Dodavatel: Honeywell Chemicals
Chlorbenzen, extra dry over Molecular Sieve 99.5%, AcroSeal®
Dodavatel: Thermo Fisher Scientific
Izobutanol 99+% for HPLC
Dodavatel: Thermo Fisher Scientific
Aceton 99.8% for HPLC
Dodavatel: Thermo Fisher Scientific