KLERCIDE 70/30 DENATURED ETHANOL 5L 1 * 4 KS
Katalogové číslo SHIE3078220
Dodavatel: ECOLAB CONTAMINATION CONTROL
KLERCIDE 70/30 DENATURED ETHANOL 5L 1 * 4 KS
Katalogové číslo SHIE3078220
Dodavatel: ECOLAB CONTAMINATION CONTROL
Frequently Bought Together
2-Propanol ≥99.5% (by GC), EMPLURA®, Supelco®
Dodavatel: Merck
1,3-Dioxolan ≥99.0% stabilised, GPR RECTAPUR® for synthesis
Dodavatel: VWR Chemicals
Formamid, EMSURE® p.a., Supelco®
Dodavatel: Merck
2-Etoxyethanol ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Pyrolidin for synthesis, Sigma-Aldrich®
Dodavatel: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
1,4-Dioxan, anhydrous ≥99.8%
Dodavatel: Thermo Fisher Scientific
Voda for TOC, Supelco®
Dodavatel: Merck
Pyridin, extra dry over Molecular Sieve 99.5%, AcroSeal®
Dodavatel: Thermo Fisher Scientific