KLERCIDE LOW RESIDUE QUAT 1L WFI STERILE 1 * 6 KS
Katalogové číslo SHIE3084120
Dodavatel: ECOLAB CONTAMINATION CONTROL
KLERCIDE LOW RESIDUE QUAT 1L WFI STERILE 1 * 6 KS
Katalogové číslo SHIE3084120
Dodavatel: ECOLAB CONTAMINATION CONTROL
Frequently Bought Together
Aceton ≥99%, TECHNICAL
Dodavatel: VWR Chemicals
Methanol ≥99.9% (by GC), HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Dodavatel: VWR Chemicals
2-Propanol ≥99.7%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Aceton, EMPARTA® ACS analytical reagent, Supelco®
Dodavatel: Merck
N,N-Dimethylformamid ≥99.9%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Etylenglykol ≥98%, TECHNICAL
Dodavatel: VWR Chemicals
Xylen (směs izomerů) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
Methanol, LiChrosolv®, hypergrade for LC-MS, for HPLC, Supelco®
Dodavatel: Merck
Glycerin ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Dodavatel: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Dodavatel: VWR Chemicals
2-Propanol, EMSURE® ACS, ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
Aceton ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
Dodavatel: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Dodavatel: VWR Chemicals
KLERCIDE SPOR.ACTIVE CHLORINE 946ML STER 1 * 6 KS
Dodavatel: ECOLAB CONTAMINATION CONTROL