Liquid soaps, Tork® Premium
Product Details & Documents
Tork® Premium soap features a reliable, drip-free system that promotes good hygiene and is labelled with the Nordic Swan.
Tork® Premium Soap Liquid Mild and Tork® Premium Soap Liquid Luxury Soft
Perfect for all normal hand washing in public areas, like hotels and restaurants
Cleans and actively conditions the skin
Liquid Mild soap: Skin friendly thanks to mild formulation
Luxury Soft soap: Silk extract for extra softness and jasmine perfume for a luxury fresh feeling.
Tork® Premium Soap Liquid Extra Mild
Perfect for frequent hand washing, especially in hygienic environments like hospitals
Perfume and colour-free, minimises the risk of allergic reactions
Tork® Premium Soap Liquid Extra Hygiene HD
Perfect to use in hygiene-sensitive environments, such as restaurants, kitchens, wellness establishments
Highly effective against bacteria and approved according to European Norms (EN 1040 and EN 1499)
Perfume and colour-free, minimises the risk of allergic reactions
Tork® Premium Soap Hair and Body
Combined soap and shampoo product for the whole body, including the hair
Perfect for showers
Mild formulation for skin and hair
Environmental license with EU Ecolabel and Nordic Ecolabel
Upozornení: *Výrobky na této stránce nespadají do oblasti působnosti směrnice EU 2019/904
Documents
Specifikace
- Popis:Luxury hair and body mini shower cream, Tork®
- Sekvence:09
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.