UTP SOLUTION (100 MM) 1 * 100 mL
Katalogové číslo ABNOU0462.100ML
Dodavatel: Abnova
Frequently Bought Together
VWR®, Láhve, širokohrdlé, se šroubovacím uzávěrem
Dodavatel: VWR Collection
Sampling scoops, LaboPlast®
Dodavatel: BURKLE
Blotting transfer papers
Dodavatel: Ahlstrom-Munksjö
Filter plates, 24-well, AcroPrep™
Dodavatel: Cytiva (Formerly Pall Lab)
AcroPrep™ 24-Well Receiver Plate
Dodavatel: Cytiva (Formerly Pall Lab)
Sample container, with screw cap
Dodavatel: DELTALAB
96-Well clear polystyrene microplates, Corning®
Dodavatel: Corning
Bioreactors, Wheaton® CELLine™
Dodavatel: DWK Life Sciences
Centrifuge tubes with flat caps, MetalFree®
Dodavatel: LABCON
Sterilin™ Samples Containers with Screw Caps, PS, 60 to 250 ml
Dodavatel: Sterilin
Centrifuge tubes, SuperClear®
Dodavatel: LABCON
Accessories and consumables for KingFisher™ Flex
Dodavatel: Thermo Fisher Scientific