[EN]DIETHYL ETHER, CHROMASOLV(R) HPLC 9 1 * 100 mL
Catalog # SIAL309966-100ML
Supplier: Merck
Frequently Bought Together
Glycerin ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
Tetrahydrofuran ≥99% stabilised, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichlormethan ≥98% stabilised, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
Aceton ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Methanol ≥98.5%, TECHNICAL
Supplier: VWR Chemicals
Aceton ≥99%, TECHNICAL
Supplier: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Voda, AnalaR NORMAPUR® ISO 3696 Grade 3 analytical reagent
Supplier: VWR Chemicals
Chlorbenzen ≥99%, GPR RECTAPUR®
Supplier: VWR Chemicals
Glycerin 85% Ph. Eur.
Supplier: VWR Chemicals
Dichlormethan ≥99.5% stabilised, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
Supplier: VWR Chemicals