[EN]MEDIUM RATCHET TUBING CLAMP 1000/PK 1 * 1.000 KS
Katalogové číslo MFLX06833-00BLK
Dodavatel: Avantor Fluid Handling
O tomto produktu
Won't rust, corrode, or distort.
- Constructed of durable Celcon plastic
Durable Celcon plastic clamp is autoclavable; won't rust, corrode, or distort. Accepts flexible tubing.
Mini clamp accept up to 1/4" OD tubing and provides on/off control. The standard clamp accepts up to 1/2" OD tubing and features twelve-position ratchet control to adjust flow from fully-open to complete-closure.
- Materiál krytu:Polyoxymethylene (POM) Acetal
- Popis:Ratchet Tubing Clamp for up to 1/2" Tubing, Acetal; 1000/pack
- Sekvence:02
- Environmentally Preferable:
- Pinch valve tube size:1/8" to 1/2" OD
- Záruka:3 Months
- Bal.:1.000
Frequently Bought Together