Orders LinkContactUsLinkComponent
Handheld refractometers, Eclipse
Handheld refractometers, Eclipse
101, Eclipse spare illuminator flaps
Katalogové číslo BELL45-006
Dodavatel:   BELLINGHAM STANLEY
Handheld refractometers, Eclipse
Katalogové číslo BELL45-006
Dodavatel:  BELLINGHAM STANLEY
Handheld refractometers, Eclipse
Handheld refractometers, Eclipse
101, Eclipse spare illuminator flaps
Přejít na:
Price
1 110,00 Kč
Přihlásit se pro dostupnost
1 * 3 KS
Product Details & Documents
Handheld refractometers use high precision clear scale, and simple zero adjustment with lock.
  • Shaded field of view for distinction
  • Anti-roll support and ergonomic design
  • Comfortable eyepiece with smooth adjustment
  • Easy to clean prism and push-on flap protection
  • Unique sample ‘dribble’ feature
Delivery information: Supplied with a soft carrying case and instructions.
Specifikace
  • Popis:
    Eclipse spare illuminator flaps
  • Sekvence:
    101