Orders LinkContactUsLinkComponent
Square HDPE Bottle
Square HDPE Bottle
03, 1000 ml, Square bottle, HDPE
Katalogové číslo SAMP8165A-1000
Dodavatel:   Sampling Systems
Square HDPE Bottle
Katalogové číslo SAMP8165A-1000
Dodavatel:  Sampling Systems
Square HDPE Bottle
Square HDPE Bottle
03, 1000 ml, Square bottle, HDPE
Přejít na:
Price
260,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Square HDPE bottles for use with a range of liquids and powders.
  • Different sizes available
  • Ribbed sides for easy handling
  • Available from stock
  • Fast delivery
This range of HDPE containers are perfect for use with a range of powders and liquids. They have a handy square space-saving shape and incorporate side ridges for easy handling when wet.
Specifikace
  • Kapacita:
    1000 ml
  • Popis:
    Square bottle
  • Materiál:
    HDPE
  • Sekvence:
    03