[EN]CALCIFEDIOL 1 * 100 mg
Katalogové číslo PHR2359-100MG
Dodavatel: Merck
Frequently Bought Together
Supelco® TraceCERT® Organic Reference Standard, Tebufenpyrad
Dodavatel: Merck
PHOSPHONIC ACID 100µG/ML IN ACETONITRILE 1 * 1 mL
Dodavatel: EHRENSTORFER
Procymidone
Dodavatel: EHRENSTORFER
Supelco® PESTANAL® Organic Reference Standard, Isoflucypram
Dodavatel: Merck
AMMONIUM CAL SOLUTION 10MG/L 1 * 500 mL
Dodavatel: YSI ENVIRONMENTAL
Standard vodivosti (25 °C; KCl)
Dodavatel: Thermo Fisher Scientific
Buffer Solutions
Dodavatel: Thermo Fisher Scientific
MCPB D6 (RING D3, METHYL D3) 100 NG/µL 1 * 1,1 mL
Dodavatel: EHRENSTORFER
CEFAZOLIN SODIUM SALT 1 * 75 mg
Dodavatel: Merck
Kyselina o-acetylsalicylová
Dodavatel: EHRENSTORFER
[EN]TOLUENE D8 1 * 0,1 g
Dodavatel: EHRENSTORFER
LYOPHILIZED EXOSOME STANDARD (U87 MG) 1 * 1 Vial
Dodavatel: Abcam
PERFLUORO-3,6-DIOXAHEPTANOIC ACID 50 µG/ML IN METHANOL:WATER 1 * 1 mL
Dodavatel: EHRENSTORFER