[EN]L-LACTIC ACID STANDARD SOLUTION 1 * 50 mL
MEGAAS-LATE
: MEGAZYME
Frequently Bought Together
2-Butoxyethanol ≥99%
: Thermo Fisher Scientific
2-Propanol 99+% for spectroscopy
: Thermo Fisher Scientific
TOLUENE (REAG. PH. EUR.) FOR ANALYSIS, A 1 * 2,5 L
: PanReac AppliChem
Toluen 99.8+% p.a.
: Thermo Fisher Scientific
Toluen ≥99.7% for spectrophotometry
: Thermo Fisher Scientific
Chloroform ≥99.5% stabilizovaný
: Honeywell Chemicals
Chloroform stabilizovaný for HPLC
: Thermo Fisher Scientific
Hexan (směs izomerů), TECHNICAL
: VWR Chemicals
Chloroform stabilizovaný, pure
: Lach-ner S.R.O.
Toluen 99.85% for HPLC
: Thermo Fisher Scientific