[EN]CALIBRATION STNDRD 0.5MG/L C NIST SU 1 * 60 mL
Katalogové číslo REGEISTOC1003
Dodavatel: REAGECON
[EN]CALIBRATION STNDRD 0.5MG/L C NIST SU 1 * 60 mL
Katalogové číslo REGEISTOC1003
Dodavatel: REAGECON
Frequently Bought Together
METHYL-4-HYDROXYBENZOATE SODIUM SALT 1 * 500 mg
Dodavatel: Merck
THIABENDAZOLE NH D6 100 NG/µL 1 * 1,1 mL
Dodavatel: EHRENSTORFER
Supelco® Organic Reference Standard, Cortisone
Dodavatel: Merck
CHLORHEXIDINEàDIACETATE 1 * 1 g
Dodavatel: Merck
[EN]MIXTURE 6 COMPONENTS 1 * 100 mL
Dodavatel: CUSTOM MADE CHEMICALS LAB
Zinc iodide and starch solution Reag. Ph. Eur. 1096502
Dodavatel: VWR Chemicals
[EN]LYCOPENE HPLC FROM TOMATO 1 * 10 mg
Dodavatel: Merck
MECOPROP D3 (RING) 100 NG/µL 1 * 1,1 mL
Dodavatel: EHRENSTORFER
BIFONAZOLE FOR SYSTEM SUITABILITY 1 * 10 mg
Dodavatel: EDQM
TESTOSTERONE PROPIONATE CERTIFIED 1 * 0,1 g
Dodavatel: EHRENSTORFER
DECOQUINATE CERTIFIED 1 * 0,1 g
Dodavatel: EHRENSTORFER
VWR®, Synthetic Sea Water Analytical Reagent manufactured in accordance with ASTM D665 / IP 135
Dodavatel: VWR Chemicals
SpecCheck secondary gel standards sets
Dodavatel: Hach
SET DIESEL OIL&MINERAL OIL(DIN H53)3X1ML 1 * 1 SET
Dodavatel: EHRENSTORFER
TEPRALOXYDIM-3-HYDROXY-GLUTARIC ACID 1 * 10 mg
Dodavatel: EHRENSTORFER
Pesticide Mixture 1
Dodavatel: EHRENSTORFER