Orders LinkContactUsLinkComponent
Pointed Scissors
Pointed Scissors
01, 150 mm
Katalogové číslo SAMPA364-150
Dodavatel:   Sampling Systems
Pointed Scissors
Katalogové číslo SAMPA364-150
Dodavatel:  Sampling Systems
Pointed Scissors
Pointed Scissors
01, 150 mm
Přejít na:
Price
268,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Surgical grade scissors for cutting.
  • Made from surgical grade 420 stainless steel
  • Great for cutting and manipulating samples
These high quality pointed scissors are made from surgical grade 420 stainless steel and great for cutting and manipulating.
Specifikace
  • Délka:
    150 mm
  • Sekvence:
    01