Orders LinkContactUsLinkComponent
Double Ended Spoons
03, Double ended spoon, 300 mm, 2/8 ml, Nerezová ocel
Katalogové číslo SAMPA104-300
Dodavatel:   Sampling Systems
Double Ended Spoons
Katalogové číslo SAMPA104-300
Dodavatel:  Sampling Systems
Double Ended Spoons
Double Ended Spoons
03, Double ended spoon, 300 mm, 2/8 ml, Nerezová ocel
Přejít na:
Price
765,00 Kč
Přihlásit se pro dostupnost
1 * 1 KS
Product Details & Documents
Double ended spoon for taking different sample sizes.
  • Made from stainless steel
  • Ideal for sampling powders and liquids
  • Various sizes available
These double ended spoons are great for taking different sample sizes using the same spoon. The lengths and volume capacities of the spoons vary allowing for flexibility in sampling both powders and liquids.
Specifikace
  • Kapacita:
    2/8 ml
  • Popis:
    Double ended spoon
  • Délka:
    300 mm
  • Materiál:
    Nerezová ocel
  • Sekvence:
    03