[EN]CM1850 HEAT DISSIPATION BLOCK FREEZE 1 * 1 KS
Katalogové číslo LEIC14047130792
Dodavatel: LEICA MICROSYSTEMS
[EN]CM1850 HEAT DISSIPATION BLOCK FREEZE 1 * 1 KS
Katalogové číslo LEIC14047130792
Dodavatel: LEICA MICROSYSTEMS
Frequently Bought Together
Gas detection tubes for hydrogen peroxide
Dodavatel: Dräger
Accessories for VWR® V-1200, UV-1600PC, UV-3100PC
Dodavatel: COLLECTION PREFIX HPLC
Accessories for titrators
Dodavatel: Mettler - Toledo
VWR® VisiScope®, Adaptér CCD pro kamery
Dodavatel: VWR Collection
Accessories for SP 100
Dodavatel: Socorex Isba
Quick connect Luer adapters, Upchurch Scientific®
Dodavatel: UPCHURCH SCIENTIFIC
Short-term tubes for accuro®
Dodavatel: Dräger
Accessories for Mettler balances
Dodavatel: Mettler - Toledo
Gas detection tubes for sulphur dioxide
Dodavatel: Dräger
pH/mV/°C meters, handheld, pH 3110 / 3310
Dodavatel: WTW
TRUB.DETEK.IMPACTOR 0,1-1MG/M3 OLEJ.MLHY 1 * 10 KS
Dodavatel: Dräger
ThermoMixer® C Thermomixer
Dodavatel: EPPENDORF
Accessories for OxiTop® and OxiTop® Control
Dodavatel: WTW
Single gas detectors, BW Clip
Dodavatel: BW TECHNOLOGIES
Melting point and boiling point tubes and capillaries
Dodavatel: BUCHI
Plastic cuvette for LICO
Dodavatel: Hach