Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
[EN]50 TABLETS TOTAL HARDNESS PHOTOMETER 1 * 50 KS
Catalog # WTIDTBSPTH50
Supplier:
WATER ID PRIMELAB
Expand image
[EN]50 TABLETS TOTAL HARDNESS PHOTOMETER 1 * 50 KS
Catalog # WTIDTBSPTH50
Supplier:
WATER ID PRIMELAB
Frequently Bought Together
[EN][EN]PONCEAU S SOLUTION 1 * 1.000 mL
Supplier: Abcam
Test strips, potassium
Supplier: MACHEREY-NAGEL
Hydroxid sodný 98%, flakes, pure
Supplier: Thermo Fisher Scientific
Test kits, chlorine (DPD), CHECKIT®
Supplier: Lovibond
Test kits, phosphate (kits and reagents) CHECKIT®
Supplier: Lovibond