[EN]50 TABLETS QAC HR PHOTOMETER 1 * 50 KS
Katalogové číslo WTIDTBSPQAC50
Dodavatel: WATER ID PRIMELAB
[EN]50 TABLETS QAC HR PHOTOMETER 1 * 50 KS
Katalogové číslo WTIDTBSPQAC50
Dodavatel: WATER ID PRIMELAB
Frequently Bought Together
Cuvettes and tubes for Picco, Multy, Nova and Prove
Dodavatel: Merck
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
Test kits, formaldehyde, Spectroquant®, Supelco®
Dodavatel: Merck
Test strips, peracetic acid, Reflectoquant®, Supelco®
Dodavatel: Merck
Formaldehyde 4% stabilised, buffered (pH 7,0 ±0,1)
Dodavatel: VWR Chemicals
Aqueous mounting medium Aquatex®, Sigma-Aldrich®
Dodavatel: Merck
Haemalum (Mayer's), Sigma-Aldrich®
Dodavatel: Merck
Schiff reagent for histology, Sigma-Aldrich®
Dodavatel: Merck
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
Dodavatel: Merck
Formalin 10%, Q Path®, buffered
Dodavatel: VWR Chemicals
METHYLENE BLUE RAL 555 AQUEOUS SOLUTION 1 * 1 L
Dodavatel: RAL
Test kits, chlorine, Spectroquant®, Supelco®
Dodavatel: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Dodavatel: VWR Chemicals
Test strips, peracetic acid, QUANTOFIX®
Dodavatel: MACHEREY-NAGEL
Reagents for copper determination using Lovibond® MD photometers
Dodavatel: Lovibond
Test kits, photometric, ammonium, LCK cuvette tests
Dodavatel: Hach