0,00 Kč
Zadání objednávky
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
[EN]50 TABLETS ORGAPHOS N 1 PHOTOMETER 1 * 50 KS
WTIDTBSPOPA50
:
WATER ID PRIMELAB
[EN]50 TABLETS ORGAPHOS N 1 PHOTOMETER 1 * 50 KS
WTIDTBSPOPA50
:
WATER ID PRIMELAB
Frequently Bought Together
Reagents for Phosphate Determination using Lovibond® MD photometers
: Lovibond
Kongo červeň indicator grade
: Thermo Fisher Scientific
Hydroxid sodný, anhydrous ≥98%, pellets, Reagent Grade
: Honeywell Chemicals
TISSUE MARKING DYE ORANGE,59ML 1 * 1 KS
: SHANDON
Nitrite VHR L, 500 ml
: Lovibond