[EN]50 TABLETS DPD N 4 PHOTOMETER 1 * 50 KS
Katalogové číslo WTIDTBSPD450
Dodavatel: WATER ID PRIMELAB
[EN]50 TABLETS DPD N 4 PHOTOMETER 1 * 50 KS
Katalogové číslo WTIDTBSPD450
Dodavatel: WATER ID PRIMELAB
Frequently Bought Together
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
Test strips, total hardness, MQuant®, Supelco®
Dodavatel: Merck
Hydroxid sodný, pellets, EMSURE® p.a., Supelco®
Dodavatel: Merck
Test kits, phosphate, Spectroquant®, Supelco®
Dodavatel: Merck
Test Strips, nitrate, MQuant®, Supelco®
Dodavatel: Merck
Mravenčan amonný ≥99%, HiPerSolv CHROMANORM® for LC-MS
Dodavatel: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Dodavatel: Merck
Hydroxid sodný, pellets, pure, Supelco®
Dodavatel: Merck
Hydroxid sodný 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Dodavatel: VWR Chemicals
Test kits, nitrate, Spectroquant®, Supelco®
Dodavatel: Merck
Test pro stanovení peroxidu, MQuant™, Supelco®
Dodavatel: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Dodavatel: VWR Chemicals
Ammonia solution 28%, AnalaR NORMAPUR® analytical reagent
Dodavatel: VWR Chemicals
Test kits, photometric, ammonium, LCK cuvette tests
Dodavatel: Hach