[EN]20G PL IRON MR 1 (285 TESTS) 1 * 20 g
Katalogové číslo WTIDPLPOW20IRONMR1
Dodavatel: WATER ID PRIMELAB
[EN]20G PL IRON MR 1 (285 TESTS) 1 * 20 g
Katalogové číslo WTIDPLPOW20IRONMR1
Dodavatel: WATER ID PRIMELAB
Frequently Bought Together
Formaldehyde 4% stabilised, buffered (pH 7,0 ±0,1)
Dodavatel: VWR Chemicals
Cuvettes and tubes for Picco, Multy, Nova and Prove
Dodavatel: Merck
Gram's decolourising solution for Gram staining, Sigma-Aldrich®
Dodavatel: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Dodavatel: Merck
Kjeldahl tablets, Kjeltabs
Dodavatel: THOMPSON & CAPPER
Paramat, Solidification point: 55 - 58 °C, in pastille form, Gurr™, without DMSO
Dodavatel: VWR Chemicals
METHYLENE BLUE RAL 555 AQUEOUS SOLUTION 1 * 1 L
Dodavatel: RAL
Test Kits, Boron, Spectroquant®, Supelco®
Dodavatel: Merck
Formalin 10%, Q Path®, buffered
Dodavatel: VWR Chemicals
[EN]SULFITE-3 250ML 424362 1 * 1 KIT
Dodavatel: Lovibond
Reagents for copper determination using Lovibond® MD photometers
Dodavatel: Lovibond
Reagents for Chlorine Analyser CL17
Dodavatel: Hach
Nitrite VHR L, 500 ml
Dodavatel: Lovibond