0,00 Kč
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Czech Republic
ContactUsLinkComponent
[EN]65ML PL MOLYBDATE 1 (162 TESTS) 1 * 65 mL
WTIDPL65MOLY1
:
WATER ID PRIMELAB
[EN]65ML PL MOLYBDATE 1 (162 TESTS) 1 * 65 mL
WTIDPL65MOLY1
:
WATER ID PRIMELAB
Frequently Bought Together
Reagents for nitrate determination using Lovibond® MD photometers
: Lovibond
Test kits, cyanide, Spectroquant®, Supelco®
: Merck
Colorimetric rapid tests, phosphate, VISOCOLOR®
: MACHEREY-NAGEL
Oxid měďnatý, wire ACS (0.65x6 mm) for elementary analysis, Supelco®
: Merck
Ammonia solution 20%, GPR RECTAPUR®
: VWR Chemicals
METHYLENE BLUE RAL 555 AQUEOUS SOLUTION 1 * 1 L
: RAL
[EN]OXALIC ACID 0.005M (0.01N) 17034 1 * 1 L
: CUSTOM MADE CHEMICALS LAB
[EN]FLUORESCENT BRIGHTENER 28 DISODIUM S 1 * 20 mL
: Merck
Reagents for Phosphate Determination using Lovibond® MD photometers
: Lovibond