[EN]65 ML PL ISOTHIAZOLINONE 4 (108 TEST 1 * 65 mL
Katalogové číslo WTIDPL65ISOZ4
Dodavatel: WATER ID PRIMELAB
[EN]65 ML PL ISOTHIAZOLINONE 4 (108 TEST 1 * 65 mL
Katalogové číslo WTIDPL65ISOZ4
Dodavatel: WATER ID PRIMELAB
Frequently Bought Together
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
Test kits, phosphate, Spectroquant®, Supelco®
Dodavatel: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
Dodavatel: Merck
Hydroxid sodný, pellets, EMSURE® p.a., Supelco®
Dodavatel: Merck
Kyselina trifluoroctová ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Dodavatel: VWR Chemicals
Test pro stanovení peroxidu, MQuant™, Supelco®
Dodavatel: Merck
Hydroxid sodný 98.5-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Dodavatel: VWR Chemicals
Test kits, COD cell test, Spectroquant®, Supelco®
Dodavatel: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
Dodavatel: VWR Chemicals
Test kits, ammonium, Spectroquant®, Supelco®
Dodavatel: Merck
pH indicator strips, MQuant®, Supelco®
Dodavatel: Merck
Kyselina mravenčí ≥99%, HiPerSolv CHROMANORM® for LC-MS
Dodavatel: VWR Chemicals
Test kits, photometric, COD, LCK cuvette tests
Dodavatel: Hach