[EN]30ML PL NICKEL HR 2 (150 TESTS) 1 * 30 mL
Katalogové číslo WTIDPL30NICKHR2
Dodavatel: WATER ID PRIMELAB
[EN]30ML PL NICKEL HR 2 (150 TESTS) 1 * 30 mL
Katalogové číslo WTIDPL30NICKHR2
Dodavatel: WATER ID PRIMELAB
Frequently Bought Together
EOSIN RAL 555 1 * 1 L
Dodavatel: RAL
VWR®, Testovací proužky, chlorid, Dosatest
Dodavatel: VWR Chemicals
Total Iron Test Strips, Dosatest, VWR®
Dodavatel: VWR Chemicals
Kyselina trifluoroctová ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Dodavatel: VWR Chemicals
Tablety pro přípravu pufrů podle Weise, Sigma-Aldrich®
Dodavatel: Merck
Test kits, chlorine, Spectroquant®, Supelco®
Dodavatel: Merck
Formalin 10%, Q Path®, buffered
Dodavatel: VWR Chemicals
Test kits, nitrite, Spectroquant®, Supelco®
Dodavatel: Merck
VWR® Kjeldahl Tablets
Dodavatel: VWR Chemicals
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
Dodavatel: Merck
Copper test strips, Dosatest
Dodavatel: VWR Chemicals
Kyselina mravenčí ≥99%, HiPerSolv CHROMANORM® for LC-MS
Dodavatel: VWR Chemicals
Reagents for hydrazine determination using Lovibond® MD photometers
Dodavatel: Lovibond
Test kits, photometric, Nitrate, LCK cuvette tests
Dodavatel: Hach