[EN]30 ML PL ISOTHIAZOLINONE 5 (150 TEST 1 * 65 mL
Katalogové číslo WTIDPL30ISOZ5
Dodavatel: WATER ID PRIMELAB
[EN]30 ML PL ISOTHIAZOLINONE 5 (150 TEST 1 * 65 mL
Katalogové číslo WTIDPL30ISOZ5
Dodavatel: WATER ID PRIMELAB
Frequently Bought Together
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
Hydroxid draselný 85.0-100.5%, pellets, AnalaR NORMAPUR® Reag. Ph. Eur. analytical reagent
Dodavatel: VWR Chemicals
Test pro stanovení peroxidu, MQuant™, Supelco®
Dodavatel: Merck
pH indicator strips, MQuant®, Supelco®
Dodavatel: Merck
Joint grease, K.W.S.
Dodavatel: POLLATH
Kyselina mravenčí ≥99%, HiPerSolv CHROMANORM® for LC-MS
Dodavatel: VWR Chemicals
Test kits, COD cell test, Spectroquant®, Supelco®
Dodavatel: Merck
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
Dodavatel: Merck
Test kits, nitrite, Spectroquant®, Supelco®
Dodavatel: Merck
Test indikátor pH proužky, pH-Fix
Dodavatel: MACHEREY-NAGEL
Kyselina trifluoroctová ≥99.9%, HiPerSolv CHROMANORM® for LC-MS
Dodavatel: VWR Chemicals
Hydroxid sodný, pellets, EMSURE® p.a., Supelco®
Dodavatel: Merck
Mravenčan amonný ≥99%, HiPerSolv CHROMANORM® for LC-MS
Dodavatel: VWR Chemicals
Reagents for chlorine determination using Lovibond® MD photometers
Dodavatel: Lovibond
VWR® Dosatest, pH indicator test strips, non bleeding
Dodavatel: VWR Chemicals
Chlornan sodný solution 5%, J.T.Baker®
Dodavatel: Avantor
Supelco® TraceCERT® Organic Reference Standard, Thiabendazole
Dodavatel: Merck