[EN]30ML PL DPD 3 C LIQUID (250 TESTS) 1 * 30 mL
Katalogové číslo WTIDPL30DPD3C
Dodavatel: WATER ID PRIMELAB
[EN]30ML PL DPD 3 C LIQUID (250 TESTS) 1 * 30 mL
Katalogové číslo WTIDPL30DPD3C
Dodavatel: WATER ID PRIMELAB
Frequently Bought Together
Hydroxid sodný 30% (w/w) in aqueous solution, AnalaR NORMAPUR® analytical reagent
Dodavatel: VWR Chemicals
Ammonia solution ∼32%, GPR RECTAPUR®
Dodavatel: VWR Chemicals
pH indicator strips, MQuant®, Supelco®
Dodavatel: Merck
NANOCOLOR® Photometric Tube Tests, Nitrate
Dodavatel: MACHEREY-NAGEL
ADF Volumetric Solution for analysis of animal feed (NF V18-122)
Dodavatel: VWR Chemicals
Kyselina 1-oktansulfonová sodná sůl ≥98%, HiPerSolv CHROMANORM® for HPLC
Dodavatel: VWR Chemicals
Test strips, peroxide, QUANTOFIX®
Dodavatel: MACHEREY-NAGEL
COD photometric test kits, Test’n Tube
Dodavatel: Hach
Reagents for hydrazine determination using Lovibond® MD photometers
Dodavatel: Lovibond
METHYLENE BLUE RAL 555 AQUEOUS SOL. 2.5L 1 * 2,5 L
Dodavatel: RAL
Hydroxid sodný ≥98.0% (by acidimetry), pellets ACS, ISO p.a.
Dodavatel: PanReac AppliChem
Test kits, photometric, Nitrate, LCK cuvette tests
Dodavatel: Hach
Test kits, photometric, COD, LCK cuvette tests
Dodavatel: Hach