[EN]30ML PL DPD 1 A LIQUID (125/250 TEST 1 * 30 mL
Katalogové číslo WTIDPL30DPD1A
Dodavatel: WATER ID PRIMELAB
[EN]30ML PL DPD 1 A LIQUID (125/250 TEST 1 * 30 mL
Katalogové číslo WTIDPL30DPD1A
Dodavatel: WATER ID PRIMELAB
Frequently Bought Together
Triethylamin ≥99.8%, HiPerSolv CHROMANORM® for LC-MS
Dodavatel: VWR Chemicals
Natronové vápno, AnalaR NORMAPUR® analytical reagent, with indicator
Dodavatel: VWR Chemicals
Schiff reagent for histology, Sigma-Aldrich®
Dodavatel: Merck
pH indicator strips, MQuant®, Supelco®
Dodavatel: Merck
VWR® Q Path® DC3, Decalcifier Rapid-Acting
Dodavatel: VWR Chemicals
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. p.a., Supelco®
Dodavatel: Merck
UltraClear® for histology, J.T.Baker®
Dodavatel: Avantor
Test strips, nitrate/nitrite, Dosatest
Dodavatel: VWR Chemicals
Chlornan sodný (12% Cl₂) in aqueous solution, GPR RECTAPUR®
Dodavatel: VWR Chemicals
Visual colour kits, iron, MQuant®, Supelco®
Dodavatel: Merck
Ammonia solution 25%, AnalaR NORMAPUR® analytical reagent
Dodavatel: VWR Chemicals
Test pro stanovení volného chloru, MQuant™, Supelco®
Dodavatel: Merck
Test kits, photometric, iron, LCK cuvette tests
Dodavatel: Hach
SODIUM HYDROXIDE SOLUTION 32 % W/W FOR N 1 * 5 L
Dodavatel: PanReac AppliChem
Fenolová červeň solution pH-indicator (pH 6,5 - 8,5)
Dodavatel: Hach